Learn More
Eurofins Discoverx Chemerin, 25 µg. (92-1036)
Supplier:  Eurofins Discoverx 921036
Encompass
Chemerin is a chemoattractant expressed in white adipose, liver, and lung tissues. Chemerin is an agonist for the G-protein coupled receptor CMKLR1 (ChemR23), which is expressed mainly on dendritic cells, macrophages, and some adipocytes. Also an agonist for GPR1. Provided in a non-glycosylated protein form. Provided as 25 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9581