Learn More
Medchemexpress LLC Chemerin-9 (149-157) (TFA) | 98.2% | 1177.18 | 1 ML
SDP
Catalog No. 5000424298
Encompass_Preferred
Chemerin-9 (149-157) TFA is a potent agonist of chemokine-like receptor 1 (CMKLR1) with anti-inflammatory activity. It stimulates the phosphorylation of Akt and ERK, and also induces ROS production. This compound has been shown to ameliorate Aβ1-42-induced memory impairment and regulates immune responses, adipocyte differentiation, and glucose metabolism.
  • Potent agonist of chemokine-like receptor 1 (CMKLR1)
  • Exhibits anti-inflammatory activity
  • Stimulates phosphorylation of Akt and ERK, and induces ROS production
  • Ameliorates Aβ1-42-induced memory impairment
  • Regulates immune responses, adipocyte differentiation, and glucose metabolism
  • Demonstrates in vitro stimulation of migration in cardiac fibroblasts
  • Alleviates glucose intolerance and IR in PDM mice in vivo
  • Provides anti-inflammatory and anti-angiogenic effects in ApoE-/- mice in vivo
Catalog No. 50-004-24298 Supplier Medchemexpress LLC Supplier No. HYP1844A10MM/1ML
Chemerin-9 (149-157) TFA is a potent agonist of chemokine-like receptor 1 (CMKLR1) with anti-inflammatory activity. It stimulates the phosphorylation of Akt and ERK, and also induces ROS production. This compound has been shown to ameliorate Aβ1-42-induced memory impairment and regulates immune responses, adipocyte differentiation, and glucose metabolism.
  • Potent agonist of chemokine-like receptor 1 (CMKLR1)
  • Exhibits anti-inflammatory activity
  • Stimulates phosphorylation of Akt and ERK, and induces ROS production
  • Ameliorates Aβ1-42-induced memory impairment
  • Regulates immune responses, adipocyte differentiation, and glucose metabolism
  • Demonstrates in vitro stimulation of migration in cardiac fibroblasts
  • Alleviates glucose intolerance and IR in PDM mice in vivo
  • Provides anti-inflammatory and anti-angiogenic effects in ApoE-/- mice in vivo