Learn More
Cayman Chemical ChemerIn-9humnacetate 500ug
SDP
Catalog No. 502909369
Encompass_Preferred
A peptide agonist of CMKLR1 and CMKLR2; binds to CMKLR1 and CMKLR2 (IC50s = 1.9 and 2.3 nM, respectively); induces calcium mobilization in HEK293T cells expressing either CMKLR1 or CMKLR2 and the G protein Gα15 at 1 µM
Catalog No. 50-290-9369 Supplier Cayman Chemical Supplier No. 36760500 UG
A peptide agonist of CMKLR1 and CMKLR2; binds to CMKLR1 and CMKLR2 (IC50s = 1.9 and 2.3 nM, respectively); induces calcium mobilization in HEK293T cells expressing either CMKLR1 or CMKLR2 and the G protein Gα15 at 1 µM