Learn More
Enzo Life Sciences Chemerin monoclonal antibody (Tigy-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804868C100
Encompass_Preferred
Clone: Tigy-1. Isotype: IgG2a. Immunogen: Recombinant rat chemerin (aa 23-157). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human, Rat. Applications: ELISA, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-0010