Learn More
CHEMetrics CHEMets Hydrogen Peroxide Test Kit Refill
SDP
Catalog No. 13299232B
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-232B 30
1 options
Catalog No. 13-299-232B Supplier CHEMetrics Supplier No. R5510
Add to Cart
Add to Cart
Provides results in two minutes or less
This product contains 30 self-filling CHEMets ampules to refill kit K-5510 for the analysis of hydrogen peroxide. R-5510 is compatible with C-5001 and C-5510 comparators.
Specifications
Analytes Hydrogen Peroxide
For Use With (Equipment) Hydrogen peroxide testing kit
Includes 30 self-filling ampoules
No. of Tests 30
Range 0 to 0.8 ppm and 1 to 10 ppm
Test Method Ferric Thiocyanate
Test Time <1 min.
Type CHEMets Hydrogen Peroxide Refiil
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Warning! Prop 65