Learn More
CHEMetrics Filming Amines Comparator
SDP
Catalog No. 13299220C
Change view
Click to view available options
Range:
0 to 1 ppm
Catalog No. Range
13-299-220C 0 to 1 ppm
1 options
Catalog No. 13-299-220C Supplier CHEMetrics Supplier No. C1001
Add to Cart
Add to Cart
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
C-1001 comparator is compatible with K-1001 and R-1000 products. Color standards are yellow in color and cover a range of 0-1 ppm in the following increments: 0, 0.05, 0.10, 0.15, 0.25, 0.50, 0.75, 1.0 ppm
Specifications
Analytes Filming Amines
For Use With (Equipment) Filming amine CHEMets kit
Range 0 to 1 ppm
Test Method Methyl Orange
Type Filming Amines Comparator