Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics CHEMets Nitrate Visual Water Test Kit
SDP

Catalog No. 13299345
Change view
Click to view available options
Range:
0 to 4.5 ppm as N
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-345 0 to 4.5 ppm as N
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-345 Supplier CHEMetrics Supplier No. K6904

missing translation for 'validMainframeMsg'

Provides results in two minutes or less

Provide results in two minutes or less

Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Comparator, 25 mL sample cup, reaction tube with lid and instructions.

Specifications

Analytes Nitrate
Includes Plastic case, Refill, Comparator, 25 mL sample cup, reaction tube with lid and instructions
No. of Tests 30
Range 0 to 4.5 ppm as N
Test Method Cadmium Reduction
Test Time 15 min.
Type CHEMets Visual Water Testing Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Warning! Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.