Learn More
CHEMetrics CHEMets Silica Visual Water Test Kit
SDP
Catalog No. 13299257
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-257 30
1 options
Catalog No. 13-299-257 Supplier CHEMetrics Supplier No. K9010
Add to Cart
Add to Cart
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
The Silica Test Kit (K-9010) comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, Neutralizer Solution, Activator Solution, 25 mL sample cup, sample cup top and instructions
Specifications
Analytes Silica
Includes Plastic case, Refill, Low and High Range Comparators, 25 mL sample cup, sample cup top, Neutralizer solution, Activator solution, instructions
No. of Tests 30
Range 0 to 1 ppm and 1 to 10 ppm
Test Method Heteropoly Blue
Test Time 2 min.
Type CHEMets Visual Water Testing Kit