Learn More
CHEMetrics CHEMets Detergents (Anionic Surfactants, MBAS) Visual Water Test Kit
SDP
Catalog No. 13299261A
Change view
Click to view available options
No. of Tests:
20
Catalog No. No. of Tests
13-299-261A 20
1 options
Catalog No. 13-299-261A Supplier CHEMetrics Supplier No. K9400
Add to Cart
Edge
Add to Cart
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
The methylene blue active substances (MBAS) method is used in a 3-minute procedure to measure anionic detergents. The procedure features a superior extraction/sampling technique that eliminates several steps required in other test procedures and provides increase sensitivity.
Specifications
Analytes Detergents, Anionic Surfactants, MBAS
Includes Comes in a cardboard box and contains everything needed to perform 20 tests: Refill, Comparator, reaction tube with lid, tip breaking tool and instructions
No. of Tests 20
Range 0 to 3 ppm
Test Method Methylene Blue
Type CHEMets Visual Water Test Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Warning! Prop 65