Learn More
CHEMetrics CHEMets Phenol Visual Water Testing Kit
SDP
Catalog No. 13299205
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-205 30
1 options
Catalog No. 13-299-205 Supplier CHEMetrics Supplier No. K8012
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, 25 mL sample cup and instructions
Specifications
Analytes Phenol
Includes Plastic case, Refill, Low and High Range Comparators, 25 mL sample cup and instructions
No. of Tests 30
Range 0 to 1 ppm and 0 to 12 ppm
Test Method 4-Aminoantipyrine
Test Time 1 min.
Type CHEMets Visual Water Testing Kit