Learn More
CHEMetrics CHEMets Cyanide (Free) Visual Water Testing Kit
SDP
Catalog No. 23291209
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
23-291209 30
1 options
Catalog No. 23-291209 Supplier CHEMetrics Supplier No. K3810
Add to Cart
Add to Cart
Provides results in two minutes or less
Provide results in two minutes or less
Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, Neutralizer Solution, Activator Solution, 25 mL sample cup, 1.0 mL syringe & instructions
Specifications
Analytes Free Cyanide
Includes Plastic case, Refill, Low and High range comparators, Neutralizer solution, Activator solution, 25 mL sample cup, 1.0 mL syringe and instructions
No. of Tests 30
Range 0 to 0.1 ppm and 0.1 to 1 ppm
Test Method Isonicotinic-Barbituric Acid
Type CHEMets Visual Water Testing Kit