Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics COD Vials Kits
SDP

Catalog No. 13299734
Change view
Click to view available options
Range:
0 to 15,000 ppm
0 to 15,000 ppm (HR+)
0 to 150 ppm
0 to 1500 ppm
Certifications/Compliance:
Not USEPA approved
USEPA accepted
No. of Tests:
24
25
149
Includes:
150 vials and instruction book
25 vials and instruction book
98 vials and instruction book
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Certifications/Compliance No. of Tests Includes
13-299-734 0 to 15,000 ppm (HR+) Not USEPA approved 24 25 vials and instruction book
13-299-730 0 to 150 ppm USEPA accepted 24 25 vials and instruction book
13-299-229 0 to 150 ppm USEPA accepted 149 150 vials and instruction book
13-299-732 0 to 1500 ppm USEPA accepted 25 25 vials and instruction book
13-299-230 0 to 1500 ppm USEPA accepted 149 150 vials and instruction book
13-299-232 0 to 15,000 ppm Not USEPA approved - 98 vials and instruction book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-299-734 Supplier CHEMetrics Supplier No. K7370S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions

Contains premeasured reagents for accurate, economical determination of COD

  • Contains everything needed (except distilled water) to perform tests
  • Includes vials and instruction book
  • Comes in cardboard box
  • 2-hour digestion time
  • Dichromate reactor digestion

A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.

Certifications

Not USEPA Approved

Specifications

Analytes Chemical Oxygen Demand (COD)
Certifications/Compliance Not USEPA approved
EPA Approved Methods Not USEPA approved
For Use With (Application) Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
For Use With (Equipment) Digestor block as well as a photometer or spectrophotometer
Includes 25 vials and instruction book
No. of Tests 24
Range 0 to 15,000 ppm (HR+)
Test Method Dichromate Reactor Digestion
Test Time 2 hr. digestion
Type COD Vials Kit
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.