Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics Dissolved Oxygen Comparator
SDP

Catalog No. 13299226
Change view
Click to view available options
Range:
0 to 20 ppb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-226 0 to 20 ppb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-299-226 Supplier CHEMetrics Supplier No. C7511
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color standard ampoules used to quantify test results for the determination of various analytes in water

  • 0 to 20 ppb
  • Shelf Life: 1 year

Specifications

Analytes Dissolved Oxygen (DO)
pH Range 2.5 to 11.5
Range 0 to 20 ppb
Type Dissolved Oxygen Comparator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.