Learn More
AquaPhoenix Scientific™ CHEMetrics™ Phenol Activator Solution Pack

Description
- Enough reagent to perform approximately 200 tests for consistent and easy dosing
Specifications
Specifications
| pH Range | 6.5 to 8.5 |
| Volume (Metric) | 120 mL |
| For Use With (Equipment) | K-8010, K-8010D, K-8043, K-8053 Phenol Test Kits |
| Sterility | Non-sterile |
| Type | Activator Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.