Learn More
AquaPhoenix Scientific™ CHEMetrics™ Phenols CHEMets Refill Kit

Description
- Designed to deliver fast, accurate results while reducing cost and waste versus replacing a full test kit
Specifications
Specifications
| pH Range | 6.5 to 8.5 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-8010, K8010D Phenol Test Kits |
| Sterility | Non-sterile |
| Test Method | 4-Aminoantipyrine |
| Type | Phenol Refill Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.