Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Scientific™ CHEMetrics™ Phenols CHEMets Visual Test Kit
SDP

Catalog No. 13299217
Change view
Click to view available options
Range:
0 to 1, 0 to 12 ppm
0 to 300 ppm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-217 0 to 300 ppm
13-299-216 0 to 1, 0 to 12 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-299-217 Supplier AquaPhoenix Scientific™ Supplier No. K8010D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kit delivers fast, accurate phenol analysis in water for wastewater, industrial, and environmental monitoring. Self-filling ampoules, ready-to-use reagents, and 1-min. results simplify testing and provide a reliable, field-proven solution.

  • Perform 30 tests

Specifications

pH Range 6.5 to 8.5
Volume (Metric) 0.5 mL
For Use With (Application) Wastewater, Industrial and Environmental Monitoring
Includes 1 Bottle Activator Solution
Range 0 to 300 ppm
Test Method 4-Aminoantipyrine
Type Phenol Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.