Learn More
CHEMetrics CHEMets Chlorine Test Kit Refill
SDP
Catalog No. 13299508B
Change view
Click to view available options
Range:
0 to 1 ppm and 1 to 5 ppm
0.1 to 2 ppm
Test Method:
DDPD
DPD
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Test Method
13-299-508B 0.1 to 2 ppm DPD
13-299-215C 0 to 1 ppm and 1 to 5 ppm DDPD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-299-508B Supplier CHEMetrics Supplier No. R2500
Add to Cart
Edge
Add to Cart
Provides results in two minutes or less
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life
Specifications
Analytes Chlorine
For Use With (Equipment) Chlorine (free and total) testing kit
Includes 30 ampoules
Range 0.1 to 2 ppm
Test Method DPD
Test Time 2 min.
Type CHEMets Refill