Learn More
Aqua Solutions Sodium Hydroxide Solution, 50% w/w, Certified, Fisher Chemical
SDP
Catalog No. NC2198926
Encompass
The Aqua Phoenix Sciences Sodium Hypochlorite 12.5% is a high-purity sodium hypochlorite solution. It contains 12.5% available chlorine and is used as a disinfectant, bleach, and cleaning agent. This chlorine bleach solution is NSF certified for use in food processing and water treatment. It comes in 5 gallon pails and 55 gallon drums. The 12.5% concentration provides rapid disinfection and cleaning. It should be handled with care as it can irritate skin and eyes. Proper PPE should be used when handling.
This item is not returnable. View return policy
Catalog No. NC2198926 Supplier Aqua Solutions Supplier No. CMK7511
The Aqua Phoenix Sciences Sodium Hypochlorite 12.5% is a high-purity sodium hypochlorite solution. It contains 12.5% available chlorine and is used as a disinfectant, bleach, and cleaning agent. This chlorine bleach solution is NSF certified for use in food processing and water treatment. It comes in 5 gallon pails and 55 gallon drums. The 12.5% concentration provides rapid disinfection and cleaning. It should be handled with care as it can irritate skin and eyes. Proper PPE should be used when handling.