Learn More
Chemgenes Corporation ChemGenes 5'-DMT 2'-ALE Guanosine, N-DMF, 3'-CE Phosphoramidite, 2 g
SDP
Catalog No. NC3940972
Encompass
The 5'-DMT 2'-ALE Guanosine (N-DMF) 3'-CE Phosphoramidite is a specialized nucleoside derivative designed for oligonucleotide synthesis. This modified guanosine phosphoramidite features an ALE (alkoxyalkyl) modification at the 2'-position and is protected with N-DMF, facilitating efficient RNA synthesis.
  • Features 5'-DMT protection for controlled coupling
  • Incorporates 3'-CE phosphoramidite for standard synthesis protocols
  • Provides 2'-ALE modification for specific structural requirements
  • Designed for high-purity oligonucleotide synthesis applications
Catalog No. NC3940972 Supplier Chemgenes Corporation Supplier No. ANP73872G
The 5'-DMT 2'-ALE Guanosine (N-DMF) 3'-CE Phosphoramidite is a specialized nucleoside derivative designed for oligonucleotide synthesis. This modified guanosine phosphoramidite features an ALE (alkoxyalkyl) modification at the 2'-position and is protected with N-DMF, facilitating efficient RNA synthesis.
  • Features 5'-DMT protection for controlled coupling
  • Incorporates 3'-CE phosphoramidite for standard synthesis protocols
  • Provides 2'-ALE modification for specific structural requirements
  • Designed for high-purity oligonucleotide synthesis applications