Learn More
Chemgenes Corporation ChemGenes Activation Reagent, 0.25 M 5-Ethylthio Tetrazole, ACN, 450 mL
SDP
Catalog No. NC3940855
Encompass
This activation reagent consists of 0.25 M 5-ethylthio-1H-tetrazole in acetonitrile. It is designed for use as an activator in oligonucleotide synthesis processes.
  • Concentration of 0.25 M
  • Composed of 5-ethylthio tetrazole in acetonitrile
  • Suitable for use in nucleic acid synthesis
Catalog No. NC3940855 Supplier Chemgenes Corporation Supplier No. RN1466450ML
This activation reagent consists of 0.25 M 5-ethylthio-1H-tetrazole in acetonitrile. It is designed for use as an activator in oligonucleotide synthesis processes.
  • Concentration of 0.25 M
  • Composed of 5-ethylthio tetrazole in acetonitrile
  • Suitable for use in nucleic acid synthesis