Learn More
Chemgenes Corporation ChemGenes Capping Reagent, 16% N-Methylimidazole, In THF, 2 L
SDP
Catalog No. NC3940877
Encompass
This capping reagent consists of a 16% solution of N-Methylimidazole in tetrahydrofuran (THF), specifically formulated for use as an ancillary reagent in oligonucleotide synthesis processes.
  • Formulated for optimal performance in oligonucleotide synthesis applications
  • Contains 16% n-methylimidazole in tetrahydrofuran
  • Produced under an iso 9001:2015 certified quality management system
Catalog No. NC3940877 Supplier Chemgenes Corporation Supplier No. RN77762L
This capping reagent consists of a 16% solution of N-Methylimidazole in tetrahydrofuran (THF), specifically formulated for use as an ancillary reagent in oligonucleotide synthesis processes.
  • Formulated for optimal performance in oligonucleotide synthesis applications
  • Contains 16% n-methylimidazole in tetrahydrofuran
  • Produced under an iso 9001:2015 certified quality management system