Learn More
Chemgenes Corporation ChemGenes DMT Removal Reagent, 3% Dichloroacetic Acid, Dichloromethane, 2 L
SDP
Catalog No. NC3940864
Encompass
DMT Removal Reagent is a chemical solution composed of 3% Dichloroacetic acid in Dichloromethane, designed for specialized laboratory applications in synthesis and purification processes.
  • Contains 3% dichloroacetic acid
  • Formulated in dichloromethane
  • Used for dmt removal
Catalog No. NC3940864 Supplier Chemgenes Corporation Supplier No. RN14682L
DMT Removal Reagent is a chemical solution composed of 3% Dichloroacetic acid in Dichloromethane, designed for specialized laboratory applications in synthesis and purification processes.
  • Contains 3% dichloroacetic acid
  • Formulated in dichloromethane
  • Used for dmt removal