Learn More
Chemgenes Corporation ChemGenes Phosphoramidite, 5'-DMT, 2'-ALE Uridine, 3'-CE, 2 g
SDP
Catalog No. NC3940966
Encompass
This product is a 5'-DMT 2'-ALE Uridine 3'-CE Phosphoramidite, designed for use in chemical synthesis applications.
  • Uridine derivative suitable for nucleic acid synthesis
  • Contains a 5'-dimethoxytrityl (DMT) protecting group
  • Features a 3'-cyanoethyl (CE) phosphoramidite moiety
  • Designed for high-performance chemical research and oligonucleotide development
Catalog No. NC3940966 Supplier Chemgenes Corporation Supplier No. ANP73842G
This product is a 5'-DMT 2'-ALE Uridine 3'-CE Phosphoramidite, designed for use in chemical synthesis applications.
  • Uridine derivative suitable for nucleic acid synthesis
  • Contains a 5'-dimethoxytrityl (DMT) protecting group
  • Features a 3'-cyanoethyl (CE) phosphoramidite moiety
  • Designed for high-performance chemical research and oligonucleotide development