Learn More
Chemglass Life Sciences Chemglass 100 mL Jacketed Reaction Vessel, 100 mm Flat Flange, Graduated Both Sides
SDP
Catalog No. NC3862664
Encompass_Preferred
Jacketed reaction vessel designed for controlled heating and cooling of small-scale laboratory reactions. The 100 mL vessel features tool-free jacket connections and graduated markings for convenient volume indication in benchtop reactor setups.
  • Jacketed design for controlled heating and cooling of small-volume reactions.
  • Graduation scale on both sides for approximate volume measurement.
  • Tool-free flat flange inlet and outlet for easy hose attachment and removal.
  • Chemically resistant perfluoro o-rings on drain valves for improved sealing.
  • Lower drain sidearm accepts standard 1/2 in or 3/4 in compression fittings.
  • Each vessel is serial numbered for traceability.
Catalog No. NC3862664 Supplier Chemglass Life Sciences Supplier No. CG1929X07
Jacketed reaction vessel designed for controlled heating and cooling of small-scale laboratory reactions. The 100 mL vessel features tool-free jacket connections and graduated markings for convenient volume indication in benchtop reactor setups.
  • Jacketed design for controlled heating and cooling of small-volume reactions.
  • Graduation scale on both sides for approximate volume measurement.
  • Tool-free flat flange inlet and outlet for easy hose attachment and removal.
  • Chemically resistant perfluoro o-rings on drain valves for improved sealing.
  • Lower drain sidearm accepts standard 1/2 in or 3/4 in compression fittings.
  • Each vessel is serial numbered for traceability.