Learn More
Chemglass Life Sciences Chemglass 2 L Glass Bottle complete with aspirator cap for fluid aspirator systems (CLS-900-510)
SDP
Catalog No. NC3852943
Encompass
The Chemglass 2 L Glass Bottle Complete with Aspirator Cap (CLS-900-510) is a replacement reservoir designed for use with fluid aspirator systems. Supplied complete with an aspirator cap, the bottle provides a durable glass container for laboratory fluid aspiration, collection, and in-bottle decontamination workflows.
  • Capacity 2 liter.
  • Material glass.
  • Includes an aspirator cap.
  • Compatible with fluid aspirator systems.
  • Sold individually (each).
Catalog No. NC3852943 Supplier Chemglass Life Sciences Supplier No. CLS900510
The Chemglass 2 L Glass Bottle Complete with Aspirator Cap (CLS-900-510) is a replacement reservoir designed for use with fluid aspirator systems. Supplied complete with an aspirator cap, the bottle provides a durable glass container for laboratory fluid aspiration, collection, and in-bottle decontamination workflows.
  • Capacity 2 liter.
  • Material glass.
  • Includes an aspirator cap.
  • Compatible with fluid aspirator systems.
  • Sold individually (each).