Learn More
Chemglass Life Sciences Chemglass High Recovery Vial, 3.7 mL, Clear Borosilicate, GPI 13-425, 15 x 45 mm, With Open-Top Cap and PTFE-Silicone Septa
SDP
Catalog No. NC3871926
Encompass
Chemglass high recovery vials (3.7 mL) are clear, Type 1 borosilicate autosampler vials engineered with a conical bottom to minimize sample loss. They are compatible with GPI 13-425 threaded closures and are supplied with an open-top cap and PTFE-silicone septa, making them suitable for analytical workflows such as HPLC and GC.
  • Maximizes sample recovery with conical bottom design.
  • Type 1 borosilicate glass provides chemical resistance and thermal stability.
  • 3.7 mL nominal capacity with a compact 15 x 45 mm footprint.
  • Compatible with GPI 13-425 threaded caps for standard autosampler fittings.
  • Supplied with open-top cap and PTFE-silicone septa for reliable sealing.
  • Clear glass allows easy visual inspection of samples.
Catalog No. NC3871926 Supplier Chemglass Life Sciences Supplier No. CEL1409191CVR
Chemglass high recovery vials (3.7 mL) are clear, Type 1 borosilicate autosampler vials engineered with a conical bottom to minimize sample loss. They are compatible with GPI 13-425 threaded closures and are supplied with an open-top cap and PTFE-silicone septa, making them suitable for analytical workflows such as HPLC and GC.
  • Maximizes sample recovery with conical bottom design.
  • Type 1 borosilicate glass provides chemical resistance and thermal stability.
  • 3.7 mL nominal capacity with a compact 15 x 45 mm footprint.
  • Compatible with GPI 13-425 threaded caps for standard autosampler fittings.
  • Supplied with open-top cap and PTFE-silicone septa for reliable sealing.
  • Clear glass allows easy visual inspection of samples.