Learn More
Chemglass Life Sciences Chemglass Life Sciences Insulated Hose, M30 Female Ends, 3.0m, -100°C to 350°C
SDP
Catalog No. NC3998553
Encompass
This insulated steel hose is designed for connecting Huber™ circulators to Chemglass reactors.
  • Made from insulated steel.
  • Features M30 female end fittings.
  • Measures 3.0m in length.
  • Operates within a temperature range of −100°C to 350°C.
  • Designed for connecting circulators to reactors.
  • Has female end fitting for direct attachment to inlet/outlet connections.
Catalog No. NC3998553 Supplier Chemglass Life Sciences Supplier No. CG1978H128
This insulated steel hose is designed for connecting Huber™ circulators to Chemglass reactors.
  • Made from insulated steel.
  • Features M30 female end fittings.
  • Measures 3.0m in length.
  • Operates within a temperature range of −100°C to 350°C.
  • Designed for connecting circulators to reactors.
  • Has female end fitting for direct attachment to inlet/outlet connections.