Learn More
Chemglass Life Sciences Chemglass Size #5 Metal Retaining Clips for Glass Stopcock Plugs, Pack of 3
SDP
Catalog No. NC2676573
Encompass_Preferred
Size #5 metal retaining clips secure glass stopcock plugs and barrels to prevent blow-out and loose fit. Made from high carbon tempered spring steel with a green corrosive-resistant, phosphate-base finish, they help minimize freezing and provide durable spring action. Sold in packs of three for laboratory stopcock maintenance.
  • Made from high carbon tempered spring steel.
  • Green corrosive-resistant, phosphate-base finish.
  • Secures stopcock plug and barrel to prevent blow-out and loose fit.
  • Helps minimize freezing of stopcock plugs.
  • Sold in packs of 3; maximum 2 packs per order.
  • Size: #5.
Catalog No. NC2676573 Supplier Chemglass Life Sciences Supplier No. CG42315
Size #5 metal retaining clips secure glass stopcock plugs and barrels to prevent blow-out and loose fit. Made from high carbon tempered spring steel with a green corrosive-resistant, phosphate-base finish, they help minimize freezing and provide durable spring action. Sold in packs of three for laboratory stopcock maintenance.
  • Made from high carbon tempered spring steel.
  • Green corrosive-resistant, phosphate-base finish.
  • Secures stopcock plug and barrel to prevent blow-out and loose fit.
  • Helps minimize freezing of stopcock plugs.
  • Sold in packs of 3; maximum 2 packs per order.
  • Size: #5.