Learn More
Chemglass Life Sciences Chemglass O-Ring, Kalrez 4079 perfluoroelastomer, Size #116 (CG-308K-116)
SDP
Catalog No. NC3901246
Encompass_Preferred
Chemglass O-Ring manufactured from Kalrez 4079 perfluoroelastomer, size #116. Designed for demanding high-temperature and chemically aggressive sealing applications, it combines elastomeric flexibility with PTFE-like chemical resistance and is suitable for use in chemical processing, semiconductor equipment, and high-temperature reactors.
  • Perfluoroelastomer material with high chemical resistance and thermal stability.
  • Service temperatures up to 316°C long-term and up to 343°C intermittently.
  • Resistant to polar solvents, strong organic solvents, acids, and strong oxidizing agents.
  • Suitable for seals in corrosive, high-temperature process equipment and reactors.
  • Not recommended for use below -5°C.
Catalog No. NC3901246 Supplier Chemglass Life Sciences Supplier No. CG308K116
Chemglass O-Ring manufactured from Kalrez 4079 perfluoroelastomer, size #116. Designed for demanding high-temperature and chemically aggressive sealing applications, it combines elastomeric flexibility with PTFE-like chemical resistance and is suitable for use in chemical processing, semiconductor equipment, and high-temperature reactors.
  • Perfluoroelastomer material with high chemical resistance and thermal stability.
  • Service temperatures up to 316°C long-term and up to 343°C intermittently.
  • Resistant to polar solvents, strong organic solvents, acids, and strong oxidizing agents.
  • Suitable for seals in corrosive, high-temperature process equipment and reactors.
  • Not recommended for use below -5°C.