Learn More
Hoefer, Inc Chemi FX imaging unit
SDP
Catalog No. NC3655200
Encompass_Preferred
"High-quality chemi imaging and gel documentation systemChemi FX gel doc system with high resolution, cooled camera and application-controlled GeneSys software for fluorescence and chemiluminescence applications.Effective chemi imaging and gel doc systemFor a laboratory that needs hassle-free chemiluminescent detection and routine gel documentation, using the Chemi FX’s powerful GeneSys software to switch between applications is simplicity itself. Place your chemi blot in, and the system’s cooled camera captures images with a sensitivity equal to film or slide in your gels to generate quality pictures of your DNA and proteins anytime you want.Featuring a standard darkroom and a 4 million pixel image resolution, the Chemi FX camera is cooled to provide a low signal-to-noise ratio with the perfect background.HI-LED lighting options cover the full spectrum of high-intensity blue, green, red and infrared resulting in faster exposure.
Catalog No. NC3655200 Supplier Hoefer, Inc Supplier No. CHEMI FX UNIT
Add to Cart
Add to Cart
Request a Quote
"High-quality chemi imaging and gel documentation systemChemi FX gel doc system with high resolution, cooled camera and application-controlled GeneSys software for fluorescence and chemiluminescence applications.Effective chemi imaging and gel doc systemFor a laboratory that needs hassle-free chemiluminescent detection and routine gel documentation, using the Chemi FX’s powerful GeneSys software to switch between applications is simplicity itself. Place your chemi blot in, and the system’s cooled camera captures images with a sensitivity equal to film or slide in your gels to generate quality pictures of your DNA and proteins anytime you want.Featuring a standard darkroom and a 4 million pixel image resolution, the Chemi FX camera is cooled to provide a low signal-to-noise ratio with the perfect background.HI-LED lighting options cover the full spectrum of high-intensity blue, green, red and infrared resulting in faster exposure.