Learn More
Hoefer, Inc Chemi FX9 Imaging Unit
SDP
Catalog No. NC3655205
Encompass_Preferred
"The perfect gel documentation system for detailed image analysisLooking for a Western blot imaging system that gives you more control over how you detect proteins? Look no further than the Chemi FX6/FX9. This powerful system, powered by Hoefer’s GeneSys software, lets you choose from a variety of chemiluminescence and fluorescence reagents from any manufacturer so that you can detect your proteins your way. With a 6 or 9 megapixel camera and HI LED lighting options, you’ll get sensitive, publication-quality images with minimal background interference. Plus, the system has unlimited copies of GeneTools analysis software to help you maximise your data.The Chemi FX6/FX9 is the perfect system for gel documentation, imaging and analysis in your lab. The in-house designed, FREE software with unlimited licences ensures you can detect chemiluminescence effectively and efficiently.
Catalog No. NC3655205 Supplier Hoefer, Inc Supplier No. CHEMI FX9
Add to Cart
Add to Cart
Request a Quote
"The perfect gel documentation system for detailed image analysisLooking for a Western blot imaging system that gives you more control over how you detect proteins? Look no further than the Chemi FX6/FX9. This powerful system, powered by Hoefer’s GeneSys software, lets you choose from a variety of chemiluminescence and fluorescence reagents from any manufacturer so that you can detect your proteins your way. With a 6 or 9 megapixel camera and HI LED lighting options, you’ll get sensitive, publication-quality images with minimal background interference. Plus, the system has unlimited copies of GeneTools analysis software to help you maximise your data.The Chemi FX6/FX9 is the perfect system for gel documentation, imaging and analysis in your lab. The in-house designed, FREE software with unlimited licences ensures you can detect chemiluminescence effectively and efficiently.