Learn More
Lakeland Industries Chemical Apron
Catalog No. 502841129
Encompass_Preferred
Looking for next-level chemical protection? Start with Lakeland's ChemMax 2 product line. This long-sleeve chemical apron features heat-sealed seams and a Saranex 23Pm film, making this garment an ideal option for chemical mixing and handling, environmental clean-up, and hazardous material remediation and response. Offered in cases of 12, C2T730 is available in sizes LG-3X.
Catalog No. 50-284-1129 Supplier Lakeland Industries Supplier No. C2T730XL
Looking for next-level chemical protection? Start with Lakeland's ChemMax 2 product line. This long-sleeve chemical apron features heat-sealed seams and a Saranex 23Pm film, making this garment an ideal option for chemical mixing and handling, environmental clean-up, and hazardous material remediation and response. Offered in cases of 12, C2T730 is available in sizes LG-3X.