Learn More
Lakeland Industries Chemical Apron
Catalog No. 502840838
Encompass_Preferred
Lakeland's ChemMax 1 chemical apron with bound seams is ideal for general industry, environmental clean-up, and chemical handling. The full sleeve and bound seam features offer additional protection against liquid splash and spray. Sold in cases of 50, this chemical-resistant apron is offered in sizes XL-5X.
Catalog No. 50-284-0838 Supplier Lakeland Industries Supplier No. C1B657Y4X
Lakeland's ChemMax 1 chemical apron with bound seams is ideal for general industry, environmental clean-up, and chemical handling. The full sleeve and bound seam features offer additional protection against liquid splash and spray. Sold in cases of 50, this chemical-resistant apron is offered in sizes XL-5X.