Learn More
Brady™ Chemical, Biohazard & Hazardous Material Labels - Adhesive Vinyl - 3 in H x 8 in W
Shop All Brady Corporation ProductsDescription
- Permanent acrylic adhesive sticks to a wide variety of surfaces
- Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
- Up to 3 year average outdoor durability
- Service temperature of -4°F to 167°F (-20°C to 75°C)
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | OSHA 1910.1200; Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704 |
| Description | Label |
| Adhesive Type | Self-adhesive |
| Color | Black/Blue/Red/Yellow on White |
| Finish | Gloss |
| Format | Label |
| Material | Vinyl |
| Printable Sides | Single-side Printable |
| Product Type | Label |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.