Learn More
Brady™ Chemical, Biohazard & Hazardous Material Labels - Self-Sticking Polyester, Spanish
Catalog No. 19107554 Shop All Brady Corporation Products
Change view
Click to view available options
Adhesive Type:
Permanent Acrylic, Self-adhesive
Catalog No. Adhesive Type
19-107-554 Permanent Acrylic, Self-adhesive
1 options
Catalog No. 19-107-554 Supplier Brady™ Supplier No. 86180
  • $31.75
    $22.65 / Pack of 5
    Save $9.10
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Color Black on Yellow
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Languages Spanish
Quantity 5/Pk.
Shape Rectangle
Width (English) 5 in.
Width (Metric) 92.71 mm