Learn More
Brady™ Chemical, Biohazard & Hazardous Material Sign - NOTICE NON-POTABLE WATER
Catalog No. 19107745 Shop All Brady Corporation Products
Catalog No. 19-107-745 Supplier Brady™ Supplier No. 95277
  • $57.50
    $43.65 / Each of 1
    Save $13.85
    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Premium Fiberglass: Extremely durable B-120 Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Graphics are permanently embedded within the sign panel, allowing for use in the most severe environments
  • Excellent chemical and abrasion resistance
  • Ideal for both indoor and outdoor applications
  • Service temperature of -40°F to 193°F (-40°C to 90°C)
Specifications
Material Fiberglass
Mounting Type Corner Holes
Shape Rectangular
Languages English
Product Type Non-potable Water Sign
Color Black, Red on White, Blue
WARNING: Cancer - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.