Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemical Changes Activity Kit
SCI_ED

Catalog No. S67416
Change view
Click to view available options
Chemical changes lab investigation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67416 Chemical changes lab investigation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S67416 Supplier Neo/Sci Corporation Supplier No. 203263

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Part of the American Chemical Society's Kids & Chemistry series.

  • Hands-on learning for young chemistry students
  • Students experiment with their environments to better understand their world
  • Combine chemicals to see the clues chemists use to determine whether chemical reactions have occurred
  • Observe color changes, the release of gas, the formation of a precipitate and the generation of heat
  • Class size: 40 students

Specifications

For Use With (Application) To observe chemical changes by recording color changes, release of gas, formation of precipitate, generation of heat
Class Size 40 Students
Product Type Chemical Changes Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.