Learn More
Electron Microscopy Sciences Chemical Container, 120 cc, 52 mm O.D.
SDP
Catalog No. 501926898
Encompass_Preferred
Relied on to transport dust samples from the moon. Ideal for small quantities of liquids or solids. Made of injection-molded polyethylene. Screw caps feature a leakproof inner seal. Easy to clean.
Volume: 120 cc
O.D.: 52 mm
Catalog No. 50-192-6898 Supplier Electron Microscopy Sciences Supplier No. 6429944
Relied on to transport dust samples from the moon. Ideal for small quantities of liquids or solids. Made of injection-molded polyethylene. Screw caps feature a leakproof inner seal. Easy to clean.
Volume: 120 cc
O.D.: 52 mm