Learn More
Electron Microscopy Sciences Chemical Container, 30 cc, 32 mm O.D.
SDP
Catalog No. 501926896
Encompass_Preferred
Relied on to transport dust samples from the moon. Ideal for small quantities of liquids or solids. Made of injection-molded polyethylene. Screw caps feature a leakproof inner seal. Easy to clean.
Volume: 30 cc
O.D.: 32 mm
Catalog No. 50-192-6896 Supplier Electron Microscopy Sciences Supplier No. 6429942
Relied on to transport dust samples from the moon. Ideal for small quantities of liquids or solids. Made of injection-molded polyethylene. Screw caps feature a leakproof inner seal. Easy to clean.
Volume: 30 cc
O.D.: 32 mm