Learn More
Research Products International Corp Chemical Control Spill Kit, Contains Absorbent and Protective Wear packed in a 5 Gallon Bucket
SDP
Catalog No. 50476438
Encompass
Universal spill control kit is effective for acids, solvents, aqueous liquids and small mercury spills. Supplied with ten absorbent pads for easy and dust free use. Each kit can absorb up to 2.5 gallons of hazardous liquids. Materials are supplied in a sealable 5 gallon polyethylene pail with carrying handle.
Kit Contains:
Safety Goggles: 2 pairs
Chemical Barrier Gloves: 2 pairs
Disposable Neoprene Gloves: 2 pairs
Disposable Nitrile Gloves: 5 pairs
Disposable Yellow Boot Covers: 2 pairs
HazMat Absorbent Pads: 10 each 15 x 18 Inches
Mercury Sponge Absorb Jar
Disposable Plastic Apron: 5 each
Waste Disposal Bags: 5 each 33 x 39 Inches
Disposable Broom
Disposable Dust Pan
Catalog No. 50-476-438 Supplier Research Products International Corp Supplier No. 114025
Universal spill control kit is effective for acids, solvents, aqueous liquids and small mercury spills. Supplied with ten absorbent pads for easy and dust free use. Each kit can absorb up to 2.5 gallons of hazardous liquids. Materials are supplied in a sealable 5 gallon polyethylene pail with carrying handle.
Kit Contains:
Safety Goggles: 2 pairs
Chemical Barrier Gloves: 2 pairs
Disposable Neoprene Gloves: 2 pairs
Disposable Nitrile Gloves: 5 pairs
Disposable Yellow Boot Covers: 2 pairs
HazMat Absorbent Pads: 10 each 15 x 18 Inches
Mercury Sponge Absorb Jar
Disposable Plastic Apron: 5 each
Waste Disposal Bags: 5 each 33 x 39 Inches
Disposable Broom
Disposable Dust Pan