Learn More
McMaster-Carr CHEMICAL HOSE 1/2NPT 1/2ID 3/4
Catalog No. NC2224720
Encompass
Extra-Flexible High-Pressure Chemical Hosewith 316 Stainless Steel 1/2 NPT Male Fittings, 1/2" ID, 3/4" OD
This item is not returnable. View return policy
Catalog No. NC2224720 Supplier McMaster-Carr Supplier No. 54685K3136
Extra-Flexible High-Pressure Chemical Hosewith 316 Stainless Steel 1/2 NPT Male Fittings, 1/2" ID, 3/4" OD