Learn More
SPS Medical CHEMICAL INDICATOR STRIP 250/P
Catalog No. NC0288187
Encompass_Preferred
This product is intended for use as an internal chemical indicator for all packages to be sterilized in Hydrogen Peroxide sterilization process (i.e. STERRAD and V-PRO). This product does not indicate sterilization; only that the indicator has been exposed to H2O2. Provides distinct color change, turning blue when exposed to H2O2.
Catalog No. NC0288187 Supplier SPS Medical Supplier No. GPS250R
This product is intended for use as an internal chemical indicator for all packages to be sterilized in Hydrogen Peroxide sterilization process (i.e. STERRAD and V-PRO). This product does not indicate sterilization; only that the indicator has been exposed to H2O2. Provides distinct color change, turning blue when exposed to H2O2.