Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Chemical and Mechanical Erosion of Rocks
GREENER_CHOICESCI_ED

Catalog No. S42868 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42868 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.
Catalog No. S42868 Supplier Innovating Science™ Supplier No. IS8704
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.

In this kit students will learn about the rock cycle and the significance of certain types of erosion. Innovating Science™ Chemical and Mechanical Erosion of Rocks kit provides an opportunity for students to investigate acid rain and other environment

The rock cycle has been the subject of study for many years. A key component to the rock cycle is the mechanical and chemical erosion of rocks.

  • In this Chemical and mechanical Erosin of Rocks kit students will learn about the rock cycle and the significance of certain types of erosion.
  • The kit included is a demonstration that will help students visualize the freezing and thawing action of water on rocks.
  • With multiple activities, this is a kit that is sure to keep students engaged throughout the whole experiment

Kit Includes:

  • 350 g Plaster of Paris
  • 30 mL 1M Hydrochloric Acid
  • 1 Bag Limestone, 50 Pieces
  • 1 Bag Granite, 50 Pieces
  • 1 Bag Sandstone, 50 Pieces
  • 4 Plastic Cups
  • 50 Plastic Bottles with Caps
  • 2 Balloons
  • 20 Disposable pipettes
  • 50 Plastic Trays

Specifications

Certifications/Compliance Product SDSs
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Science Education
Includes 350 g Plaster of Paris, 30 mL 1M Hydrochloric Acid, 1 Bag Limestone Pieces, 50 Pieces, 1 Bag Granite Pieces, 50 Pieces, 1 Bag Sandstone Pieces, 50 Pieces, 4 Plastic Cups, 50 Plastic Bottles with Caps, 2 Balloons, 20 Disposable pipettes, 50 Plastic
Quantity 1
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 15 Lab Groups
Product Type Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.