Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemical and Mechanical Weathering of Rocks Lab Investigation
SCI_ED

Catalog No. S32687
Change view
Click to view available options
Chem and Mech Weathering Of Rock
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S32687 Chem and Mech Weathering Of Rock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S32687 Supplier Neo/Sci Corporation Supplier No. 201563

Explore how weathering affects the Earth's surface.

  • Observe the action and rate of chemical weathering processes like oxidation, carbonation, and acidity
  • Study the effect that environmental factors such as temperature and abrasion have on rocks
  • Determine which rock types are most affected and which chemicals cause the greatest impact

Key Science Curriculum Concepts:
Carbonation, chemical weathering, mechanical weathering, oxidation and rock types

Cross-Curriculum Integration:
Agriculture, Chemistry, Environmental Science and Weather

Order Info

Class size: 40 students.

Specifications

For Use With (Application) Explore how weathering affects the earth's surface
Product Type Chemical and Mechanical Weathering of Rocks Lab Investigation
Sufficient For 40 Students
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.