Learn More
Grainger Chem-Tech Chemical Metering Pump, Diaphragm, Degassing, 100 psi, 24 gpd, 1 ea
Catalog No. 502793336
Encompass_Preferred
The Prime Performance series chemical metering pump is a motorized-diaphragm pump designed for dependable and consistent chemical delivery in a compact form. Featuring a specialized degassing valve system, this pump is optimized for off-gassing chemicals such as sodium hypochlorite, making it suitable for applications in pools, spas, and water treatment. The unit is constructed with a durable PVC pump head and an industrial-grade, low-stress diaphragm for longevity in continuous duty cycles.
  • Degassing valve system with top-mounted, one-way vent assembly
  • Adjustable stroke system with locking lever for flow rate control
  • Capable of flow rates from 2.4 gpd to 24.0 gpd
  • Maximum operating pressure of 100 psi
  • Includes 16 ft of tubing and valve/strainer/injection assembly
  • Conforms to ANSI/NSF STD. 50
  • Compact, injection-molded housing with PVC head
Catalog No. 50-279-3336 Supplier Grainger Supplier No. 1AKE7
The Prime Performance series chemical metering pump is a motorized-diaphragm pump designed for dependable and consistent chemical delivery in a compact form. Featuring a specialized degassing valve system, this pump is optimized for off-gassing chemicals such as sodium hypochlorite, making it suitable for applications in pools, spas, and water treatment. The unit is constructed with a durable PVC pump head and an industrial-grade, low-stress diaphragm for longevity in continuous duty cycles.
  • Degassing valve system with top-mounted, one-way vent assembly
  • Adjustable stroke system with locking lever for flow rate control
  • Capable of flow rates from 2.4 gpd to 24.0 gpd
  • Maximum operating pressure of 100 psi
  • Includes 16 ft of tubing and valve/strainer/injection assembly
  • Conforms to ANSI/NSF STD. 50
  • Compact, injection-molded housing with PVC head