Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Docetaxel, Microtubule-Stabilizing Agent, >99% Purity, 1 mg
SDP

Catalog No. 50195874
Encompass

Docetaxel belongs to the taxane family of antitumor and antileukemic agents. It is a microtubule-stabilizing agent that acts as an inhibitor of cellular functions requiring microtubule dynamics, including cell division.

  • Acts as an inhibitor of microtubule-dependent cellular functions, such as cell division.
  • Supplied as 1 mg powder.
  • Purity: >99%.
  • Soluble in DMSO at 200 mg/mL and ethanol at 100 mg/mL.
  • Recommended for research use only.
  • Storage: store lyophilized or in solution at -20°C, desiccated.

Catalog No. 50-195-874 Supplier Cell Signaling Technology Supplier No. 9886S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Docetaxel belongs to the taxane family of antitumor and antileukemic agents. It is a microtubule-stabilizing agent that acts as an inhibitor of cellular functions requiring microtubule dynamics, including cell division.

  • Acts as an inhibitor of microtubule-dependent cellular functions, such as cell division.
  • Supplied as 1 mg powder.
  • Purity: >99%.
  • Soluble in DMSO at 200 mg/mL and ethanol at 100 mg/mL.
  • Recommended for research use only.
  • Storage: store lyophilized or in solution at -20°C, desiccated.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.