Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology U0126, Chemical Modulator, MEK1/2 Inhibitor, 5 mg
SDP

Catalog No. 50195887
Encompass

U0126 is a highly selective inhibitor of MEK1 and MEK2 that functions by binding to the kinase in a mutually exclusive manner with other inhibitors like PD98059. This compound is used in research to study the mitogen activated protein kinase cascade, which controls cell growth and differentiation. It does not inhibit other protein kinases such as c-Abl, Raf, MEKK, Erk, JNK, MKK3, MKK4/SEK1, MKK6, CDK2, or CDK4.

  • Highly selective inhibitor of MEK1 and MEK2
  • Supplied as a lyophilized beige to light brown powder
  • Molecular weight of 380.5 g/mol
  • Purity ≥98%
  • Stable for 24 months in lyophilized form when stored at -20°C
  • Compatible with DMSO or methanol for stock preparation

Catalog No. 50-195-887 Supplier Cell Signaling Technology Supplier No. 9903S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

U0126 is a highly selective inhibitor of MEK1 and MEK2 that functions by binding to the kinase in a mutually exclusive manner with other inhibitors like PD98059. This compound is used in research to study the mitogen activated protein kinase cascade, which controls cell growth and differentiation. It does not inhibit other protein kinases such as c-Abl, Raf, MEKK, Erk, JNK, MKK3, MKK4/SEK1, MKK6, CDK2, or CDK4.

  • Highly selective inhibitor of MEK1 and MEK2
  • Supplied as a lyophilized beige to light brown powder
  • Molecular weight of 380.5 g/mol
  • Purity ≥98%
  • Stable for 24 months in lyophilized form when stored at -20°C
  • Compatible with DMSO or methanol for stock preparation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.