Learn More
Grainger Oberon Company Face Shield, Polycarbonate, Chemical Resistant, Anti-Fog, 1 ea
Catalog No. 502757947
Encompass_Preferred
This chemical-resistant face shield provides reliable protection against most acids, bases, and solvents. Constructed from heat-resistant polycarbonate, the unit is designed for maximum strength and long-lasting durability. The design features an anti-fog coating and an enlarged viewing window for enhanced visibility.
  • Provides protection against chemical splashes
  • Constructed from heat-resistant polycarbonate for strength
  • Offers a 20% larger viewing area than standard shields
  • Includes soft, ratcheting headgear that conforms to the head to eliminate pinch points
  • Compliant with ANSI Z87.1+ standards
Catalog No. 50-275-7947 Supplier Grainger Supplier No. 150X39
This chemical-resistant face shield provides reliable protection against most acids, bases, and solvents. Constructed from heat-resistant polycarbonate, the unit is designed for maximum strength and long-lasting durability. The design features an anti-fog coating and an enlarged viewing window for enhanced visibility.
  • Provides protection against chemical splashes
  • Constructed from heat-resistant polycarbonate for strength
  • Offers a 20% larger viewing area than standard shields
  • Includes soft, ratcheting headgear that conforms to the head to eliminate pinch points
  • Compliant with ANSI Z87.1+ standards