Learn More
Electron Microscopy Sciences Chemical Polishing Fixture, Model 160
SDP
Catalog No. 50981004
Encompass
The Model 160 is ideal for damage free polishing of semiconductor materials with harsh chemicals and has been used extensively for polishing GaAs with bromine-methanol solutions. The Model 160 is made of "PTFE" and is designed to hold crystal wafers pt o 2 1/8" diameter while they are being polished with very corrosive chemicals. The sample is held in place by vacuum and the sample is polished by rubbing the exposed sample against a suitable polishing cloth soaked with the chemical solution.
Catalog No. 50-981-004 Supplier Electron Microscopy Sciences Supplier No. 5016460
The Model 160 is ideal for damage free polishing of semiconductor materials with harsh chemicals and has been used extensively for polishing GaAs with bromine-methanol solutions. The Model 160 is made of "PTFE" and is designed to hold crystal wafers pt o 2 1/8" diameter while they are being polished with very corrosive chemicals. The sample is held in place by vacuum and the sample is polished by rubbing the exposed sample against a suitable polishing cloth soaked with the chemical solution.