Learn More
Lanx Chemical Protective Boot Liner
Catalog No. 191300236D
Catalog No. 19-130-0236D Supplier Lanx Supplier No. CPUSOCKSXL
Add to Cart
Add to Cart
Durable with outstanding vapor, aerosol, and fire protection
  • Unique stretch charteristics offer form-fitting comfort and minimizes sizing tariff
  • Air permeable to reduce heat stress via evaporative cooling
  • Mid-calf design integreates with most footwear
Order Info
Can be laundered multiple times on gentle cyle in non-phosphate detergents and tumble dried on low.
Compliance
Lanx Type I fabric meets U.S. military specification MIL-U-44435.
Specifications
Certifications/Compliance MIL-U-44435
For Use With (Application) Reduce heat stress
Material Composite Fabric